Merck
  • Casticin inhibits the epithelial-mesenchymal transition in ovarian carcinoma via the hedgehog signaling pathway.
Casticin inhibits the epithelial-mesenchymal transition in ovarian carcinoma via the hedgehog signaling pathway.
Oncology letters (2018-03-16)
Jing Zhang, Yinghong Cui, Shuwen Sun, Jianguo Cao, Xiaoling Fang
ABSTRACT
Casticin inhibits migration, invasion and induced apoptosis in numerous cancer cells; however, the Hedgehog (Hh) signaling pathway is a key factor in the epithelial-mesenchymal transition (EMT). The present study aimed to assess whether casticin affects the expression of members of the Hh signaling pathway and EMT effectors in ovarian carcinoma. The ovarian cancer SKOV3 cell line was incubated in the presence of various concentrations of casticin or cyclopamine. Next, the expression levels of the main Hh signaling effector glioma-associated oncogene-1 (Gli-1) and EMT-associated factors [Twist-related protein 1 (Twist1), E-cadherin and N-cadherin] were determined by western blotting and reverse transcription-quantitative polymerase chain reaction. Cell proliferation and growth were assessed using MTT and soft agar assays; cell migration and invasion was evaluated using an in vitro migration assay and a transwell invasion assay, respectively. Compared with control group values, Gli-1, Twist1 and N-cadherin expression levels were reduced, whereas E-cadherin levels were increased in the casticin- and cyclopamine-treated groups. Incubation with casticin or cyclopamine resulted in markedly reduced SKOV3 cell viability, migration and invasion, in a dose-dependent manner. To the best of our knowledge, the findings of the present study indicated for first time that casticin may inhibit EMT via Hh signaling in vitro, reducing the migratory ability of ovarian cancer cells.
Sigma-Aldrich
Monoclonal Anti-N-Cadherin antibody produced in mouse, clone GC-4, ascites fluid
Sigma-Aldrich
Anti-E-cadherin antibody produced in rabbit, affinity isolated antibody