
Trandolapril Related Compound D

Synonym(s):
Ethyl (2S)-2-[(3S,5aS,9aR,10aS)-3-methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl]-4-phenylbutanoate, Trandolapril diketopiperazine
Empirical Formula (Hill Notation):
C24H32N2O4
CAS Number:
Molecular Weight:
412.52
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2