Skip to Content
Merck
Sodium aminosalicylate dihydrate for equipment qualification European Pharmacopoeia (EP) Reference Standard

Sodium aminosalicylate dihydrate for equipment qualification

Sodium aminosalicylate dihydrate for equipment qualification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Sodium 4-aminosalicylate dihydrate, 4-Amino-2-hydroxybenzoic acid sodium salt, 4-Amino-salicylic acid sodium salt
Empirical Formula (Hill Notation):
C7H6NNaO3 · 2H2O
CAS Number:
Molecular Weight:
211.15
EC Number:

Products

Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing