
Netilmicin for peak identification

Synonym(s):
Netilmicin sulfate salt, 1-N-Ethylsisomicin
Linear Formula:
C21H41N5O7 · 5/2H2SO4
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing