
Grubbs Catalyst® M102 ChemBeads

Synonym(s):
Grubbs Catalyst® M102
Empirical Formula (Hill Notation):
C43H72Cl2P2Ru
CAS Number:
Molecular Weight:
822.96
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing