Skip to Content
Merck
Butylbenzene analytical standard

Butylbenzene

Butylbenzene analytical standard
Synonym(s):
1-Phenylbutane
Linear Formula:
C6H5(CH2)3CH3
CAS Number:
Molecular Weight:
134.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing