
Brivaracetam CRS

Synonym(s):
Brivaracetam, (αS,4R)-α-Ethyl-2-oxo-4-propyl-1-pyrrolidineacetamide, (2S)-2-[(4R)-2-Oxo-4-propylpyrrolidin-1-yl]butanamide, (S)-2-((R)-2-Oxo-4-propylpyrrolidin-1-yl)butanamide, UCB 34714, UCB-34714, UCB34714
Empirical Formula (Hill Notation):
C11H20N2O2
CAS Number:
Molecular Weight:
212.29
Products
Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing