
4-(tert-Butoxycarbonyl)phenylboronic acid, pinacol ester

Empirical Formula (Hill Notation):
C17H25BO4
CAS Number:
Molecular Weight:
304.19
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing