Merck
LC/MS Analysis of Erythromycin Analogs on Ascentis® Express HILIC
LC/MS Analysis of Erythromycin Analogs on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
Erythromycin ethyl succinate
Erythromycin
tested according to Ph. Eur.
Erythromycin C
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Ascentis Express HILlC, 15 cm x 4.6 mm I.D., 2.7 μm particles (53981-U)
mobile phase
[A] 100 mM ammonium formate, pH 3.0 (titrated with formic acid); [B] acetonitrile; (10:90, A:B)
flow rate
1.0 mL/min
pressure
1247 psi (86 bar)
column temp.
35 °C
detector
ESI(+); SIR
injection
5 μL
Description
Analysis Note
This application shows the suitability of Ascentis Express Silica columns for the analysis of erythromycin analogs in aqueous normal phase/HILIC mode of chromatography. Other relatively hydrophobic aminoglycosides have also been shown to separate under these conditions.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany