Merck
  • 5,5'-Dibromo-bis(3'-indolyl)methane induces Kruppel-like factor 4 and p21 in colon cancer cells.
5,5'-Dibromo-bis(3'-indolyl)methane induces Kruppel-like factor 4 and p21 in colon cancer cells.
Molecular cancer therapeutics (2008-07-23)
Sung Dae Cho, Sudhakar Chintharlapalli, Maen Abdelrahim, Sabitha Papineni, Shengxi Liu, Jingjing Guo, Ping Lei, Ala Abudayyeh, Stephen Safe
ABSTRACT
Bis(3'-indolyl)methane (DIM) is a metabolite of the phytochemical indole-3-carbinol, and both compounds exhibit a broad spectrum of anticancer activities. We have developed a series of synthetic symmetrical ring-substituted DIM analogues, including 5,5'-dibromoDIM, which are more potent than DIM as inhibitors of cancer cell and tumor growth. In colon cancer cells, 5,5'-dibromoDIM decreased cell proliferation and inhibited G(0)-G(1)- to S-phase progression, and this was accompanied by induction of the cyclin-dependent kinase inhibitor p21 in HT-29 and RKO colon cancer cells. Mechanistic studies showed that induction of p21 in both RKO (p53 wild-type) and HT-29 (p53 mutant) cells by 5,5'-dibromoDIM was Krüppel-like factor 4 (KLF4) dependent, and induction of p53 in RKO cells was also KLF4 dependent. Analysis of the p21 promoter in p53-dependent RKO cells showed that 5,5'-dibromoDIM activated p21 gene expression through the proximal GC-rich sites 1 and 2, and chromatin immunoprecipitation assays showed that KLF4 and p53 bound to this region of the promoter, whereas in HT-29 cells unidentified upstream cis-elements were required for induction of p21. 5,5'-DibromoDIM (30 mg/kg/d) also inhibited tumor growth and induced p21 in athymic nude mice bearing RKO cells as xenografts, showing that ring-substituted DIM such as 5,5'-dibromoDIM represent a novel class of mechanism-based drugs for clinical treatment of colon cancer.
Sigma-Aldrich
5-Bromoindole-3-carboxaldehyde, 98%
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).