Merck
  • Adenylyl cyclase regulates heavy metal sensitivity, bikaverin production and plant tissue colonization in Fusarium proliferatum.
Adenylyl cyclase regulates heavy metal sensitivity, bikaverin production and plant tissue colonization in Fusarium proliferatum.
Journal of basic microbiology (2010-01-19)
Gábor Kohut, Brigitta Oláh, Attila L Adám, Jorge García-Martínez, László Hornok
ABSTRACT
A homologue of the adenylyl cyclase (AC) gene of Neurospora crassa, named Fpacy1 was cloned from the genomic library of Fusarium proliferatum ITEM 2287 by screening the library with a DNA fragment amplified by using PCR primers designed from conserved sequences of the catalytic domain of AC genes from other fungi. The deduced FPACY1 protein had 53-77% identity with the AC proteins of other fungi. DeltaFpacy1 mutants obtained by targeted gene disruption showed retarded vegetative growth, increased conidiation and delayed conidial germination. Colonization capability of the mutants, assessed on maize seedlings and tomato fruits also was adversely affected. In sexual crosses the AC mutants retained full male fertility, but their female fertility decreased significantly. Disruption of Fpacy1 abolished vegetative self-incompatibility, suggesting that the AC gene is involved in multiple developmental processes related to vegetative growth, as well as sexual and parasexual events. The elevated thermo- and H(2)O(2)-tolerance of the DeltaFpacy1 mutants was coupled to an increased sensitivity towards Cd and Cu, indicating that the cAMP signaling pathway may have both negative and positive regulatory roles on the stress response mechanisms of fungal cells. When grown under nitrogen limitation conditions, the DeltaFpacy1 mutants produced an average of approximately 274 microg g(-1) bikaverin, whereas only traces of this metabolite was detected in the wild type. This finding provides further evidence of the role of the cAMP-PKA pathway in regulating bikaverin production.
Sigma-Aldrich
Bikaverin, from Fusarium subglutinans, ≥98% (HPLC)