Merck
  • Discriminative stimulus properties of beta-phenylethylamine, deuterated beta-phenylethylamine, phenylethanolamine and some metabolites of phenylethylamine in rodents.
Discriminative stimulus properties of beta-phenylethylamine, deuterated beta-phenylethylamine, phenylethanolamine and some metabolites of phenylethylamine in rodents.
Pharmacology, biochemistry, and behavior (1986-06-01)
D Reid, A J Goudie
ABSTRACT
The discriminative stimulus (cue) properties of phenylethylamine (PEA) were analysed in rodents in a conventional two lever FR10 operant drug discrimination task. Rats trained to discriminate phenylethylamine at 30 mg/kg showed complete dose-related generalization to PEA and to two potential PEA metabolites: phenylethanolamine (PEOH) and N-Methyl PEA (NMPEA). Only partial (50%) generalization was seen with N-Methylphenylethanolamine (NMPEOH), another potential PEA metabolite. The specificity of PEA's action as a discriminative stimulus was demonstrated by the finding that fenfluramine, a substituted phenylethylamine, failed to generalize to PEA even at high doses with marked behavioural effects which are known to have discriminative stimulus properties themselves. These data suggest that NMPEA and PEOH may be functionally important active metabolites of PEA, particularly if the major pathway of PEA metabolism to phenylacetic acid under the influence of MAO Type B is for any reason impaired. A long acting deuterium substituted form of PEA (alpha, alpha, d2 PEA), which is resistant to metabolism by MAO, produced complete dose-related generalization to the PEA cue but was more potent than PEA, due presumably to its resistance to metabolism by MAO. Deuterated PEA may therefore be a useful agent to use in future studies of the PEA cue, because the discriminability of PEA itself appears to be low due to its very rapid metabolism in vivo.
Sigma-Aldrich
N-Methyl-phenethylamine, 99%