Merck
  • Synthesis, structure and biological activity of nickel(II) complexes of 5-methyl 2-furfural thiosemicarbazone.
Synthesis, structure and biological activity of nickel(II) complexes of 5-methyl 2-furfural thiosemicarbazone.
Journal of inorganic biochemistry (2001-09-22)
E M Jouad, G Larcher, M Allain, A Riou, G M Bouet, M A Khan, X D Thanh
ABSTRACT
5-Methyl 2-furfuraldehyde thiosemicarbazone (M5HFTSC) with nickel(II) leads to three types of complexes: [Ni(M5HFTSC)(2)X(2)], [Ni(M5FTSC)(2)] and [Ni(M5FTSC)(2)] x 2DMF. In the first type the ligand remains in thione form, while in the two other, the anionic thiolato form is involved. The species [Ni(M5HFTSC)(2)X(2)] has been characterized spectroscopically. The structures of [Ni(M5FTSC)(2)] x 2DMF and [Ni(M5FTSC)(2)] have been solved using X-ray diffraction. Biological studies of [Ni(M5HFTSC)(2)Cl(2)] have been carried out in vitro for antifungal activity on human pathogenic fungi, Aspergillus fumigatus and Candida albicans, and in vivo for toxicity on mice. The results are compared to those of the ligand, the metal salt and a similar copper complex [Cu(M5HFTSC)Cl(2)].
Sigma-Aldrich
5-Methylfurfural, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
5-Methylfurfural, ReagentPlus®, 99%
Pricing and availability is not currently available.