Merck
  • Cytoplasmic cleavage of IMPA1 3' UTR is necessary for maintaining axon integrity.
Cytoplasmic cleavage of IMPA1 3' UTR is necessary for maintaining axon integrity.
Cell reports (2021-02-25)
Catia Andreassi, Raphaëlle Luisier, Hamish Crerar, Marousa Darsinou, Sasja Blokzijl-Franke, Tchern Lenn, Nicholas M Luscombe, Giovanni Cuda, Marco Gaspari, Adolfo Saiardi, Antonella Riccio
ABSTRACT
The 3' untranslated regions (3' UTRs) of messenger RNAs (mRNAs) are non-coding sequences involved in many aspects of mRNA metabolism, including intracellular localization and translation. Incorrect processing and delivery of mRNA cause severe developmental defects and have been implicated in many neurological disorders. Here, we use deep sequencing to show that in sympathetic neuron axons, the 3' UTRs of many transcripts undergo cleavage, generating isoforms that express the coding sequence with a short 3' UTR and stable 3' UTR-derived fragments of unknown function. Cleavage of the long 3' UTR of Inositol Monophosphatase 1 (IMPA1) mediated by a protein complex containing the endonuclease argonaute 2 (Ago2) generates a translatable isoform that is necessary for maintaining the integrity of sympathetic neuron axons. Thus, our study provides a mechanism of mRNA metabolism that simultaneously regulates local protein synthesis and generates an additional class of 3' UTR-derived RNAs.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Upf1 Antibody, from rabbit