Merck
  • Mycophenolic acid (MPA) modulates host cellular autophagy progression in sub genomic dengue virus-2 replicon cells.
Mycophenolic acid (MPA) modulates host cellular autophagy progression in sub genomic dengue virus-2 replicon cells.
Microbial pathogenesis (2019-09-29)
Nageswar Reddy Manchala, Ranjeet Dungdung, Pankaj Trivedi, Unnikrishnan Unniyampurath, Rajendra Pilankatta
ABSTRACT
Cellular autophagy (Macrophagy) is a self-degradative process, executed through the network of autophagy associated genes (ATGs) encoded proteins. Both in vitro and in vivo studies suggest that dengue virus (DENV) induces autophagy and supports the viral genome replication and translation. Therefore, the cellular autophagy induced by dengue virus can be a good target for antiviral drug development. The action of mycophenolic acid (MPA), a specific inhibitor of DENV replication, was investigated in the stable BHK-21/DENV2 replicon cells. The inhibition was mediated by enhanced degradation of autophagic substrates in stable BHK-21/DENV2 replicon cells as evidenced by a decrease in lapidated LC3 (LC3II) and p62 expression in the presence of MPA. In contrast, the results indicated that four gene sets, namely Transmembrane protein 74 (TMEM74), Unc-51-like kinase 2 (ULK2), Cathepsin D (CTSD) and Estrogen receptor 1 (ESR1) were upregulated in stable BHK-21/DENV2 replicon cells, due to the sustained dynamic replication of DENV2 genome. These ATGs involved in the pre-autophagosomal structure (PAS) formation, were suppressed in the presence MPA. Instead, MPA induced the expression of different set of autophagy genes such as ATG4, AKT1, APP, ATG16L1, ATG16L2, B2M and HPRT1. An enzyme involved in the nucleotide salvage pathway, HPRT1, was highly expressed in the presence of MPA. The study shows that DENV2 replication is dependent on PAS formation and is inhibited in the presence of MPA by enhancing the degradation of autophagic substrates and suppression of PAS formation. This study provides impetus in designing MPA analogues to effectively inhibit dengue viral replication.
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution