Merck
  • Safety, Tumor Reduction, and Clinical Impact of Zika Virus Injection in Dogs with Advanced-Stage Brain Tumors.
Safety, Tumor Reduction, and Clinical Impact of Zika Virus Injection in Dogs with Advanced-Stage Brain Tumors.
Molecular therapy : the journal of the American Society of Gene Therapy (2020-03-30)
Carolini Kaid, Raquel Azevedo Dos Santos Madi, Renato Astray, Ernesto Goulart, Luiz Carlos Caires-Junior, Thiago Giove Mitsugi, Ana Carolina Ramos Moreno, Maria Fernanda Castro-Amarante, Lennon Ramos Pereira, Bruna Felício Milazzotto Maldonado Porchia, Thais Oliveira de Andrade, Vivian Landini, Daniel Soares Sanches, Carolina Gonçalves Pires, Rubens Koji Oliveira Tanioka, Marcia C L Pereira, Igor Neves Barbosa, Cristina O Massoco, Luís Carlos de Souza Ferreira, Oswaldo Keith Okamoto, Mayana Zatz
ABSTRACT
Malignant brain tumors are among the most aggressive cancers with poor prognosis and no effective treatment. Recently, we reported the oncolytic potential of Zika virus infecting and destroying the human central nervous system (CNS) tumors in vitro and in immunodeficient mice model. However, translating this approach to humans requires pre-clinical trials in another immunocompetent animal model. Here, we analyzed the safety of Brazilian Zika virus (ZIKVBR) intrathecal injections in three dogs bearing spontaneous CNS tumors aiming an anti-tumoral therapy. We further assessed some aspects of the innate immune and inflammatory response that triggers the anti-tumoral response observed during the ZIKVBR administration in vivo and in vitro. For the first time, we showed that there were no negative clinical side effects following ZIKVBR CNS injections in dogs, confirming the safety of the procedure. Furthermore, the intrathecal ZIKVBR injections reduced tumor size in immunocompetent dogs bearing spontaneous intracranial tumors, improved their neurological clinical symptoms significantly, and extended their survival by inducing the destruction specifically of tumor cells, sparing normal neurons, and activating an immune response. These results open new perspectives for upcoming virotherapy using ZIKV to destroy and induce an anti-tumoral immune response in CNS tumors for which there are currently no effective treatments.
Product Description
Millipore
MILLIPLEX® Canine Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.