Merck
  • Supramolecular solvents formation in aqueous solutions containing primary amine and monoterpenoid compound: Liquid phase microextraction of sulfonamides.
Supramolecular solvents formation in aqueous solutions containing primary amine and monoterpenoid compound: Liquid phase microextraction of sulfonamides.
Talanta (2020-05-28)
Polina Bogdanova, Aleksei Pochivalov, Christina Vakh, Andrey Bulatov
ABSTRACT
A phenomenon of supramolecular solvents formation in aqueous solutions containing primary amine and monoterpenoid compound is presented for the first time. A novel supramolecular solvent-based liquid phase microextraction was developed. In an aqueous phase primary amine formed isotropic solution due to formation of amphiphile supramolecular assembly. The phase separation was achieved by a coacervation process. The monoterpenoid phenol (thymol) acted as a coacervation agent and induced spontaneous in situ formation of tiny supramolecular solvent droplets. The applicability of the proposed microextraction procedure was demonstrated for the separation of sulfonamides from biological fluids (human plasma and serum). An obtained composition of supramolecular solvent (1-decylamine, thymol, water) promoted satisfactory extraction of polar analytes from aqueous phase. The calibration graphs were linear over the concentration ranges of 0.06-50Ā mgĀ L-1 for sulfamethoxazole, sulfamethazine and sulfapyridine. The limit of detection calculated from the blank tests based on 3Ļƒ was 0.02Ā mgĀ L-1 for all analytes. The total analysis time was 15Ā min.
Sigma-Aldrich
Heptylamine, 99%