Merck
  • Snail1-dependent p53 repression regulates expansion and activity of tumour-initiating cells in breast cancer.
Snail1-dependent p53 repression regulates expansion and activity of tumour-initiating cells in breast cancer.
Nature cell biology (2016-10-28)
Ting Ni, Xiao-Yan Li, Na Lu, Teng An, Zhi-Ping Liu, Rong Fu, Wen-Cong Lv, Yi-Wei Zhang, Xiao-Jun Xu, R Grant Rowe, Yong-Shun Lin, Amanda Scherer, Tamar Feinberg, Xiao-Qi Zheng, Bao-An Chen, X Shirley Liu, Qing-Long Guo, Zhao-Qiu Wu, Stephen J Weiss
ABSTRACT
The zinc-finger transcription factor Snail1 is inappropriately expressed in breast cancer and associated with poor prognosis. While interrogating human databases, we uncovered marked decreases in relapse-free survival of breast cancer patients expressing high Snail1 levels in tandem with wild-type, but not mutant, p53. Using a Snail1 conditional knockout model of mouse breast cancer that maintains wild-type p53, we find that Snail1 plays an essential role in tumour progression by controlling the expansion and activity of tumour-initiating cells in preneoplastic glands and established tumours, whereas it is not required for normal mammary development. Growth and survival of preneoplastic as well as neoplastic mammary epithelial cells is dependent on the formation of a Snail1/HDAC1/p53 tri-molecular complex that deacetylates active p53, thereby promoting its proteasomal degradation. Our findings identify Snail1 as a molecular bypass that suppresses the anti-proliferative and pro-apoptotic effects exerted by wild-type p53 in breast cancer.
Sigma-Aldrich
Anti-ATM phosphoSer1981 Antibody, clone 10H11.E12, clone 10H11.E12, Chemicon®, from mouse
Sigma-Aldrich
Anti-MDM2 Antibody, clone 3G9, clone 3G9, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)