Merck
  • Flexibility and sorption selectivity in rigid metal-organic frameworks: the impact of ether-functionalised linkers.
Flexibility and sorption selectivity in rigid metal-organic frameworks: the impact of ether-functionalised linkers.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-12-09)
Sebastian Henke, Rochus Schmid, Jan-Dierk Grunwaldt, Roland A Fischer
ABSTRACT
The functionalisation of well-known rigid metal-organic frameworks (namely, [Zn(4)O(bdc)(3)](n), MOF-5, IRMOF-1 and [Zn(2)(bdc)(2)(dabco)](n); bdc = 1,4-benzene dicarboxylate, dabco = diazabicyclo[2.2.2]octane) with additional alkyl ether groups of the type -O-(CH(2))(n)-O-CH(3) (n = 2-4) initiates unexpected structural flexibility, as well as high sorption selectivity towards CO(2) over N(2) and CH(4) in the porous materials. These novel materials respond to the presence/absence of guest molecules with structural transformations. We found that the chain length of the alkyl ether groups and the substitution pattern of the bdc-type linker have a major impact on structural flexibility and sorption selectivity. Remarkably, our results show that a high crystalline order of the activated material is not a prerequisite to achieve significant porosity and high sorption selectivity.
Sigma-Aldrich
2-Hydroxyterephthalic acid, 97%