Merck
  • Novel prognostic protein markers of resectable pancreatic cancer identified by coupled shotgun and targeted proteomics using formalin-fixed paraffin-embedded tissues.
Novel prognostic protein markers of resectable pancreatic cancer identified by coupled shotgun and targeted proteomics using formalin-fixed paraffin-embedded tissues.
International journal of cancer (2012-08-24)
Tatsuyuki Takadate, Tohru Onogawa, Tetsuya Fukuda, Fuyuhiko Motoi, Takashi Suzuki, Kiyonaga Fujii, Makoto Kihara, Sayaka Mikami, Yasuhiko Bando, Shimpei Maeda, Kazuyuki Ishida, Takashi Minowa, Nobutaka Hanagata, Hideo Ohtsuka, Yu Katayose, Shinichi Egawa, Toshihide Nishimura, Michiaki Unno
ABSTRACT
Pancreatic cancer is among the most lethal malignancies worldwide. We aimed to identify novel prognostic markers by applying mass spectrometry (MS)-based proteomic analysis to formalin-fixed paraffin-embedded (FFPE) tissues. Resectable, node positive pancreatic ductal adenocarcinoma (PDAC) with poor (n = 4) and better (n = 4) outcomes, based on survival duration, with essentially the same clinicopathological backgrounds, and noncancerous pancreatic ducts (n = 5) were analyzed. Cancerous and noncancerous cells collected from FFPE tissue sections by laser microdissection (LMD) were processed for liquid chromatography (LC)-tandem MS (MS/MS). Candidate proteins were identified by semiquantitative comparison and then analyzed quantitatively using selected reaction monitoring (SRM)-based MS. To confirm the associations between candidate proteins and outcomes, we immunohistochemically analyzed a cohort of 87 cases. In result, totally 1,229 proteins were identified and 170 were selected as candidate proteins for SRM-based targeted proteomics. Fourteen proteins overexpressed in cancerous as compared to noncancerous tissue showed different expressions in the poor and better outcome groups. Among these proteins, we found that three novel proteins ECH1, OLFM4 and STML2 were overexpressed in poor group than in better group, and that one known protein GTR1 was expressed reciprocally. Kaplan-Meier analysis showed high expressions of all four proteins to correlate with significantly worse overall survival (p < 0.05). In conclusion, we identified four proteins as candidates of prognostic marker of PDAC. The combination of shotgun proteomics verified by SRM and validated by immunohistochemistry resulted in the prognostic marker discovery that will contribute the understanding of PDAC biology and therapeutic development.
Sigma-Aldrich
Anti-ECH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution