
Succinylacetone

Synonym(s):
4,6-Dioxoheptanoic acid, Succinylacetone
Linear Formula:
CH3COCH2COCH2CH2CO2H
CAS Number:
Molecular Weight:
158.15
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing