![Lansoprazole for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/167/504/c3b6df28-3dbf-4d18-8d5a-67bdb0b5140a/640/c3b6df28-3dbf-4d18-8d5a-67bdb0b5140a.png)
Lansoprazole for peak identification
![Lansoprazole for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/167/504/c3b6df28-3dbf-4d18-8d5a-67bdb0b5140a/640/c3b6df28-3dbf-4d18-8d5a-67bdb0b5140a.png)
Synonym(s):
Lansoprazole
Empirical Formula (Hill Notation):
C16H14F3N3O2S
CAS Number:
Molecular Weight:
369.36
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing