
Fosfomycin calcium

Synonym(s):
Phosphomycin calcium salt, (−)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, Fosfomycin calcium salt, Phosphonomycin
Linear Formula:
C3H5O4PCa
CAS Number:
Molecular Weight:
176.12
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing