Skip to Content
Merck

ERE-042

Explosives Stock Standard

2000 μg/mL each component in ethyl acetate, ampule of 1.2 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

2-Amino-4,6-Dinitrotoluene , 4-Amino-2,6-Dinitrotoluene , 3,5-Dinitroaniline , 1,3-Dinitrobenzene , 2,4-Dinitrotoluene , 2,6-Dinitrotoluene , Nitrobenzene , 2-Nitrotoluene , 3-Nitrotoluene , 4-Nitrotoluene , RDX , Tetryl , 1,3,5-Trinitrobenzene , 2,4,6-Trinitrotoluene (TNT)

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

2000 μg/mL each component in ethyl acetate

application(s)

environmental

format

multi-component solution

storage temp.

−20°C

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Analyte

Description

  • 1,3,5-Trinitrobenzene

  • 1,3-Dinitrobenzene

  • 2,4-Dinitrotoluene

  • 2,6-Dinitrotoluene

  • 2-Amino-4,6-dinitrotoluene

  • 3,5-Dinitroaniline

  • 3-Nitrotoluene

  • 4-Amino-2,6-dinitrotoluene

  • 4-Nitrotoluene

  • Nitrobenzene

  • RDX

  • Tetryl

See All (12)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

26.6 °F - closed cup

flash_point_c

-3 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library