Skip to Content
Merck
All Photos(2)

Documents

GF60321079

Yttrium

powder, 500 max. part. size (micron), weight 100 g, purity 99.9%

Synonym(s):

Y 006010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
MDL number:
UNSPSC Code:
12141617
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

powder

manufacturer/tradename

Goodfellow 603-210-79

resistivity

57 μΩ-cm, 20°C

weight

100 g

max. part. size

500 (micron)

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service