Merck
  • Post-translational modifications of Medicago truncatula glutathione peroxidase 1 induced by nitric oxide.
Post-translational modifications of Medicago truncatula glutathione peroxidase 1 induced by nitric oxide.
Nitric oxide : biology and chemistry (2017-02-15)
Claude Castella, Ioanna Mirtziou, Aurélie Seassau, Alexandre Boscari, Françoise Montrichard, Kalliopi Papadopoulou, Nicolas Rouhier, Alain Puppo, Renaud Brouquisse
ABSTRACT
Plant glutathione peroxidases (Gpx) catalyse the reduction of various peroxides, such as hydrogen peroxide (H
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
Enterokinase from bovine intestine, powder