Merck
Development of a catalytic aromatic decarboxylation reaction.
Organic letters (2007-06-05)
Joshua S Dickstein, Carol A Mulrooney, Erin M O'Brien, Barbara J Morgan, Marisa C Kozlowski
ABSTRACT
A palladium-catalyzed aromatic decarboxylation reaction has been developed. With electron-rich aromatic acids, the reaction proceeds efficiently under fairly mild conditions in good yields. The method was useful with complex functionalized substrates containing hindered carboxylic acids.
Sigma-Aldrich
Palladium(II) trifluoroacetate, 97%