Merck
  • Synthesis, crystal structure, vibration spectral, and DFT studies of 4-aminoantipyrine and its derivatives.
Synthesis, crystal structure, vibration spectral, and DFT studies of 4-aminoantipyrine and its derivatives.
Molecules (Basel, Switzerland) (2013-01-25)
Yi Li, Yuanyuan Liu, Haowei Wang, Xiaohui Xiong, Ping Wei, Fangshi Li
ABSTRACT
Three compounds derived from 4-aminoantipyrine (AA) were synthesized and their structures confirmed by melting point, elemental analysis, FT-IR, and 1H-NMR. The molecular structures of the four compounds were characterized by single-crystal X-ray diffraction and calculated by using the density functional theory (DFT) method with 6-31G (d) basis set. The calculated molecular geometries and the vibration frequencies of the AA derivatives in the ground state have been compared with the experimental data. The results show that the optimized geometries can reproduce well the crystal structural parameters, and the theoretical vibration frequencies show good agreement with the experimental data, although the experimental data are different from the theoretical ones due to the intermolecular forces. Besides, the molecular electrostatic potential (MEP) and the frontier molecular orbital (FMO) analysis of the compounds were investigated by theoretical calculations.
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%