Merck
  • Diamines interfere with the transport of L-ornithine in Ehrlich-cell plasma-membrane vesicles.
Diamines interfere with the transport of L-ornithine in Ehrlich-cell plasma-membrane vesicles.
The Biochemical journal (1991-12-15)
M A Medina, J L Urdiales, J M Mates, I Núñez de Castro, F Sánchez-Jiménez
ABSTRACT
1. L-Ornithine transport by plasma-membrane vesicles isolated from Ehrlich cells is Na(+)-independent and shows a saturable and a diffusional component. 2. Putrescine, histamine, 5-hydroxytryptamine and 2,3-diaminopropane at 55 microM concentration significantly inhibit 0.5 mM-L-ornithine transport at least for the first 10 min of incubation. 3. There is a trans-stimulatory effect of putrescine on L-ornithine transport.
Sigma-Aldrich
(S)-(−)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
1,2-Diaminopropane, 99%
Sigma-Aldrich
(R)-(+)-1,2-Diaminopropane dihydrochloride, 99%