Merck
  • Validated gas chromatographic-mass spectrometric assay for determination of the antifreezes ethylene glycol and diethylene glycol in human plasma after microwave-assisted pivalylation.
Validated gas chromatographic-mass spectrometric assay for determination of the antifreezes ethylene glycol and diethylene glycol in human plasma after microwave-assisted pivalylation.
Journal of chromatography. B, Biomedical sciences and applications (2001-05-08)
H H Maurer, F T Peters, L D Paul, T Kraemer
ABSTRACT
A gas chromatographic-mass spectrometric assay is described for identification and quantification of the antifreezes ethylene glycol (EG) and diethylene glycol (DEG) in plasma for early diagnosis of a glycol intoxication. After addition of 1,3-propanediol as internal standard, the plasma sample was deproteinized by acetone and an aliquot of the supernatant was evaporated followed by microwave-assisted pivalylation. After gas chromatographic separation, the glycols were first identified by comparison of the full mass spectra with reference spectra and then quantified. The quantification has been validated according to the criteria established by the Journal of Chromatography B. The assay was found to be selective. The calibration curves for EG and DEG were linear from 0.1 g/l to 1.0 g/l. The limit of detection for EG and DEG was 0.01 g/l and the limit of quantification for both was 0.1 g/l. The absolute recoveries were 50 and 65% for the low quality control samples and 51 and 73% for the high quality control samples of EG and DEG, respectively. Intra- and inter-day accuracy and precision were inside the required limits. The glycols in frozen plasma samples were stable for more than 6 months. The method was successfully applied to several authentic plasma samples from patients intoxicated with glycols. It has also been suitable for analysis of EG and DEG in urine.
Product Description
Supelco
Ethylene glycol, analytical standard
Pricing and availability is not currently available.