Merck
  • One-year efficacy and safety of routine prasugrel in patients with acute coronary syndromes treated with percutaneous coronary intervention: results of the prospective rijnmond collective cardiology research study.
One-year efficacy and safety of routine prasugrel in patients with acute coronary syndromes treated with percutaneous coronary intervention: results of the prospective rijnmond collective cardiology research study.
Netherlands heart journal : monthly journal of the Netherlands Society of Cardiology and the Netherlands Heart Foundation (2018-06-23)
T Yetgin, E Boersma, P C Smits, A G de Vries, E Huijskens, F Zijlstra, M M J M van der Linden, R J M van Geuns
ABSTRACT
To investigate 1‑year outcomes with routine prasugrel treatment after acute coronary syndrome (ACS) in a large-scale registry. The Rijnmond Collective Cardiology Research registry is a prospective, observational study that enrolled 4,258 consecutive ACS patients treated with percutaneous coronary intervention (PCI) with 1‑year follow-up. Patients received prasugrel as first-choice antiplatelet agent, except for increased bleeding risk patients in which clopidogrel was recommended. Events were validated by an independent clinical endpoint committee. A total number of 2,677 patients received prasugrel at discharge after the index event. Eighty-one percent of the target population was on prasugrel treatment at hospital discharge. At 1 year, the primary endpoint, a composite of all-cause mortality and myocardial infarction, occurred in 2.4% of patients receiving prasugrel. All-cause mortality occurred in 1.0%, myocardial infarction in 1.5%, target-vessel revascularisation in 3.1%, stent thrombosis in 0.6%, and stroke in 0.5% of the patients treated with prasugrel. Thrombolysis in Myocardial Infarction defined major bleeding episodes not related to coronary artery bypass grafting were observed in 1.4% of patients receiving prasugrel. In routine practice, a tailored approach of prasugrel prescription in ACS patients undergoing PCI, resulted in low ischaemic and low bleeding rates up to 1 year post PCI.
Sigma-Aldrich
Prasugrel, ≥98% (HPLC)