
Ubiquinol solution

Empirical Formula (Hill Notation):
C59H92O4
CAS Number:
Molecular Weight:
865.36
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing