Skip to Content
Merck
Heptadecanoyl chloride 98%

Heptadecanoyl chloride

Heptadecanoyl chloride 98%
Synonym(s):
Margaroyl chloride
Linear Formula:
CH3(CH2)15COCl
CAS Number:
Molecular Weight:
288.90

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing