Skip to Content
Merck
(−)-Epinephrine (+)-bitartrate salt solid

(−)-Epinephrine (+)-bitartrate salt

(−)-Epinephrine (+)-bitartrate salt solid
Synonym(s):
L-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, L-Epinephrine D-hydrogentartrate
Empirical Formula (Hill Notation):
C9H13NO3 · C4H6O6
CAS Number:
Molecular Weight:
333.29
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing