Skip to Content
Merck
Azacitidine Pharmaceutical Secondary Standard; Certified Reference Material

Azacitidine

Azacitidine Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
5-Azacytidine, 4-Amino-1-(β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, Ladakamycin
Empirical Formula (Hill Notation):
C8H12N4O5
CAS Number:
Molecular Weight:
244.20
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing