Merck
  • Uniform electroactive fibre-like micelle nanowires for organic electronics.
Uniform electroactive fibre-like micelle nanowires for organic electronics.
Nature communications (2017-06-27)
Xiaoyu Li, Piotr J Wolanin, Liam R MacFarlane, Robert L Harniman, Jieshu Qian, Oliver E C Gould, Thomas G Dane, John Rudin, Martin J Cryan, Thomas Schmaltz, Holger Frauenrath, Mitchell A Winnik, Charl F J Faul, Ian Manners
ABSTRACT
Micelles formed by the self-assembly of block copolymers in selective solvents have attracted widespread attention and have uses in a wide variety of fields, whereas applications based on their electronic properties are virtually unexplored. Herein we describe studies of solution-processable, low-dispersity, electroactive fibre-like micelles of controlled length from π-conjugated diblock copolymers containing a crystalline regioregular poly(3-hexylthiophene) core and a solubilizing, amorphous regiosymmetric poly(3-hexylthiophene) or polystyrene corona. Tunnelling atomic force microscopy measurements demonstrate that the individual fibres exhibit appreciable conductivity. The fibres were subsequently incorporated as the active layer in field-effect transistors. The resulting charge carrier mobility strongly depends on both the degree of polymerization of the core-forming block and the fibre length, and is independent of corona composition. The use of uniform, colloidally stable electroactive fibre-like micelles based on common π-conjugated block copolymers highlights their significant potential to provide fundamental insight into charge carrier processes in devices, and to enable future electronic applications.
Sigma-Aldrich
[1,3-Bis(diphenylphosphino)propane]dichloronickel(II)
Sigma-Aldrich
2,5-Dibromo-3-hexylthiophene, 97%
Sigma-Aldrich
Trichloro(octadecyl)silane, ≥90%