Merck
  • Mitochondrially Targeted Nanoparticles Based on α-TOS for the Selective Cancer Treatment.
Mitochondrially Targeted Nanoparticles Based on α-TOS for the Selective Cancer Treatment.
Macromolecular bioscience (2015-12-04)
Raquel Palao-Suay, Laura Rodrigáñez, María Rosa Aguilar, Carolina Sánchez-Rodríguez, Francisco Parra, Mar Fernández, Juan Parra, Juan Riestra-Ayora, Ricardo Sanz-Fernández, Julio San Román
ABSTRACT
The aim of this work is the preparation of an active nanovehicle for the effective administration of α-tocopheryl succinate (α-TOS). α-TOS is loaded in the core of nanoparticles (NPs) based on amphiphilic pseudo-block copolymers of N-vinyl pyrrolidone and a methacrylic derivative of α-TOS. These well-defined spherical NPs have sizes below 165 nm and high encapsulation efficiencies. In vitro activity of NPs is tested in hypopharynx squamous carcinoma (FaDu) cells and nonmalignant epithelial cells, demonstrating that the presence of additional α-TOS significantly enhances its antiproliferative activity; however, a range of selective concentrations is observed. These NPs induce apoptosis of FaDu cells by activating the mitochondria death pathway (via caspase-9). Both loaded and unloaded NPs act via complex II and produce high levels of reactive oxygen species that trigger apoptosis. Additionally, these NPs effectively suppress the vascular endothelial growth factor (VEGF) expression of human umbilical vein endothelial cells (HUVECs). These results open the possibility to use this promising nanoformulation as an α-TOS delivery system for the effective cancer treatment, effectively resolving the current limitations of free α-TOS administration.
Sigma-Aldrich
N749 Black Dye