Merck
  • Organocatalytic asymmetric direct vinylogous aldol reactions of γ-crotonolactone with aromatic aldehydes.
Organocatalytic asymmetric direct vinylogous aldol reactions of γ-crotonolactone with aromatic aldehydes.
Chemical communications (Cambridge, England) (2010-11-12)
Sunil V Pansare, Eldho K Paul
ABSTRACT
The direct aldol reaction of γ-crotonolactone and various aromatic aldehydes is catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts to provide diastereomerically and enantiomerically enriched 5-substituted 2(5H) furanones (γ-butenolides). The reaction is a simple alternative to the classical vinylogous aldol reaction of silyloxy furans.
Sigma-Aldrich
2(5H)-Furanone, 98%