Merck
  • A fully validated method for the simultaneous determination of 11 antidepressant drugs in whole blood by gas chromatography-mass spectrometry.
A fully validated method for the simultaneous determination of 11 antidepressant drugs in whole blood by gas chromatography-mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2012-06-05)
Ioannis Papoutsis, Alaa Khraiwesh, Panagiota Nikolaou, Constantinos Pistos, Chara Spiliopoulou, Sotirios Athanaselis
ABSTRACT
Antidepressant drugs are widely used for the treatment of depression and other psychiatric disorders and as a result they are involved in numerous clinical and forensic cases. The aim of this study was the development, optimization and validation of a simple, specific and sensitive GC/MS method for the simultaneous determination of 11 antidepressant drugs and 4 of their metabolites (amitriptyline, citalopram, clomipramine, fluoxetine, fluvoxamine, maprotiline, desmethyl-maprotiline, mirtazapine, desmethyl-mirtazapine, nortriptyline, paroxetine, sertraline, desmethyl-sertraline, venlafaxine and desmethyl-venlafaxine) in whole blood. The combination of solid-phase extraction with derivatization using heptafluorobutyric anhydride efficiently reduced matrix effect and improved sensitivity of the method. In this assay, protriptyline was used as internal standard. Absolute recovery values for all analytes were ranged from 79.2 to 102.6%. LODs and LOQs were found to be between 0.30-1.50 μg/L and 1.00-5.00 μg/L, respectively. The calibration curves were linear (R(2)≥0.990) within the range of 5.00-1000 μg/L for all analytes. Accuracy expressed as the % E(r) was found to be between -12.3 and 12.2%. Precision expressed as the % RSD was found to be less than 11.7% for all antidepressants. The developed method proved to be suitable for routine work and it was used to successfully analyze more than 2500 clinical and forensic blood samples.
Product Description
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
Heptafluorobutyric anhydride, ≥98%