Merck
  • Effector memory alphabeta T lymphocytes can express FcgammaRIIIa and mediate antibody-dependent cellular cytotoxicity.
Effector memory alphabeta T lymphocytes can express FcgammaRIIIa and mediate antibody-dependent cellular cytotoxicity.
Journal of immunology (Baltimore, Md. : 1950) (2008-04-09)
Béatrice Clémenceau, Régine Vivien, Mathilde Berthomé, Nelly Robillard, Richard Garand, Géraldine Gallot, Solène Vollant, Henri Vié
ABSTRACT
Human memory T cells are comprised of distinct populations with different homing potential and effector functions: central memory T cells that mount recall responses to Ags in secondary lymphoid organs, and effector memory T cells that confer immediate protection in peripheral tissues. In the present study we demonstrate that a proportion of effector memory T cells express FcgammaRIIIa (CD16), are perforin positive, and directly mediate Ab-dependent cytotoxicity ex vivo. This particular alphabeta T lymphocyte subset has the morphology of large granular lymphocytes, increases proportionately in vivo during reactive lymphocytosis, and can be detected in vitro among EBV-specific T lymphocytes after stimulation with EBV Ags. Consequently, during a normal immune response, amplification of these effector memory T lymphocytes that are capable of Ab-dependent cytotoxicity may have beneficial or harmful consequences depending on the presence of pathogen- or tissue-specific Abs, respectively.
Product Description
Millipore
Protein-Concentrate Kit (Micro), Protein-Concentrate Kit (Micro) is suitable for concentrating proteins for running gels, raising antibodies, protein purification, protein assays & other applications.