Merck
  • Novel Surface-Modified Bilosomes as Functional and Biocompatible Nanocarriers of Hybrid Compounds.
Novel Surface-Modified Bilosomes as Functional and Biocompatible Nanocarriers of Hybrid Compounds.
Nanomaterials (Basel, Switzerland) (2020-12-17)
Ewelina Waglewska, Agata Pucek-Kaczmarek, Urszula Bazylińska
ABSTRACT
In the present contribution, we demonstrate a new approach for functionalization of colloidal nanomaterial consisting of phosphatidylcholine/cholesterol-based vesicular systems modified by FDA-approved biocompatible components, i.e., sodium cholate hydrate acting as a biosurfactant and Pluronic P123-a symmetric triblock copolymer comprising poly(ethylene oxide) (PEO) and poly(propylene oxide) (PPO) blocks Eight novel bilosome formulations were prepared using the thin-film hydration method followed by sonication and extrusion in combination with homogenization technique. The optimization studies involving the influence of the preparation technique on the nanocarrier size (dynamic light scattering), charge (electrophoretic light scattering), morphology (transmission electron microscopy) and kinetic stability (backscattering profiles) revealed the most promising candidate for the co-loading of model active compounds of various solubility; namely, hydrophilic methylene blue and hydrophobic curcumin. The studies of the hybrid cargo encapsulation efficiency (UV-Vis spectroscopy) exhibited significant potential of the formulated bilosomes in further biomedical and pharmaceutical applications, including drug delivery, anticancer treatment or diagnostics.
Sigma-Aldrich
Sodium cholate hydrate, ≥97.0% (dried material, NT)
Pricing and availability is not currently available.