Merck
  • Detection of Human Cytomegalovirus Proteins in Paraffin-Embedded Breast Cancer Tissue Specimens-A Novel, Automated Immunohistochemical Staining Protocol.
Detection of Human Cytomegalovirus Proteins in Paraffin-Embedded Breast Cancer Tissue Specimens-A Novel, Automated Immunohistochemical Staining Protocol.
Microorganisms (2021-06-03)
Joel Touma, Yan Liu, Afsar Rahbar, Mattia Russel Pantalone, Nerea Martin Almazan, Katja Vetvik, Cecilia Söderberg-Nauclér, Jürgen Geisler, Torill Sauer
ABSTRACT
Emerging evidence supports a significant association between human cytomegalovirus (HCMV) and human malignancies, suggesting HCMV as a human oncomodulatory virus. HCMV gene products are found in >90% of breast cancer tumors and seem to be correlated with more aggressive disease. The definitive diagnosis of HCMV relies on identification of virus inclusions and/or viral proteins by different techniques including immunohistochemical staining. In order to reduce biases and improve clinical value of HCMV diagnostics in oncological pathology, automation of the procedure is needed and this was the purpose of this study. Tumor specimens from 115 patients treated for primary breast cancer at Akershus University Hospital in Norway were available for the validation of the staining method in this retrospective study. We demonstrate that our method is highly sensitive and delivers excellent reproducibility for staining of HCMV late antigen (LA), which makes this method useful for future routine diagnostics and scientific applications.
Sigma-Aldrich
Anti-Cytomegalovirus Antibody, clone 8B1.2, clone 8B1.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cytomegalovirus Antibody, late Antigen, clone 1G5.2, clone 1G5.2, Chemicon®, from mouse