Merck
  • Stem cell proliferation patterns as an alternative for in vivo prediction and discrimination of carcinogenic compounds.
Stem cell proliferation patterns as an alternative for in vivo prediction and discrimination of carcinogenic compounds.
Scientific reports (2017-05-04)
An-Sofie Stevens, Maxime Willems, Michelle Plusquin, Jan-Pieter Ploem, Ellen Winckelmans, Tom Artois, Karen Smeets
ABSTRACT
One of the major challenges in the development of alternative carcinogenicity assays is the prediction of non-genotoxic carcinogens. The variety of non-genotoxic cancer pathways complicates the search for reliable parameters expressing their carcinogenicity. As non-genotoxic and genotoxic carcinogens have different cancer risks, the objective of this study was to develop a concept for an in vivo test, based on flatworm stem cell dynamics, to detect and classify carcinogenic compounds. Our methodology entails an exposure to carcinogenic compounds during the animal's regeneration process, which revealed differences in proliferative responses between non-genotoxic and genotoxic carcinogens during the initial stages of the regeneration process. A proof of concept was obtained after an extensive study of proliferation dynamics of a genotoxic and a non-genotoxic compound. A pilot validation with a limited set of compounds showed that the proposed concept not only enabled a simple prediction of genotoxic and non-genotoxic carcinogens, but also had the power to discriminate between both. We further optimized this discrimination by combining stem cell proliferation responses with a phenotypic screening and by using specific knockdowns. In the future, more compounds will be tested to further validate and prove this concept.
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Phenobarbital sodium salt
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, biotin conjugate, Upstate®, from rabbit