
Morantel hydrogen tartrate

Synonym(s):
Morantel (+)-tartrate salt, 1,4,5,6-Tetrahydro-1-methyl-2-(2-[3-methyl-2-thienyl]ethenyl)pyrimidine
Empirical Formula (Hill Notation):
C12H16N2S · C4H6O6
CAS Number:
Molecular Weight:
370.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing