
Fluticasone propionate Related Compound D

Synonym(s):
6α,9-Difluoro-17-[(methylsulfanyl)carbonyl]-11β-hydroxy-16α-methyl-3-oxoandrosta-1,4-dien-17α-yle propanoate, S-Methyl 6α, 9α-difluoro-11β-hydroxy-16α-methyl-3-oxo-17α-propionyloxyandrosta-1,4-diene-17β-carbothioate, Ticabesone propionate
Empirical Formula (Hill Notation):
C25H32F2O5S
CAS Number:
Molecular Weight:
482.58
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing