
Flutamide for system suitability

Synonym(s):
Flutamide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Empirical Formula (Hill Notation):
C11H11F3N2O3
CAS Number:
Molecular Weight:
276.21
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing