
Diclofenac Impurity B

Synonym(s):
2-[(2,6-Dichlorophenyl)amino]benzaldehyde, 2-(2,6-Dichloroanilino)benzaldehyde, N-(2,6-Dichlorophenyl)anthranilaldehyde
Empirical Formula (Hill Notation):
C13H9Cl2NO
CAS Number:
Molecular Weight:
266.12
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing