
Cyclopropyldiphenylsulfonium tetrafluoroborate

Linear Formula:
C3H5S(C6H5)2BF4
CAS Number:
Molecular Weight:
314.15
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing