
Anti-PTPRF antibody produced in rabbit

Synonym(s):
Anti-LAR, Anti-Leukocyte antigen-related tyrosine phosphatase, Anti-Tyrosine-protein phosphatase receptor type F
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1