Skip to Content
Merck
All Photos(1)

Documents

Detroit 562

87042205, human pharynx, Heteroploid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Detroit 562, 87042205

biological source

human pharynx

growth mode

Adherent

karyotype

2n = 46

morphology

Heteroploid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian pharynx carcinoma

Cell Line Description

Derived from pleural fluid of an adult female with primary carcinoma of the pharynx. The cells have the type B glucose-6-phosphate dehydrogenase.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 12
D16S539: 11
D5S818: 11,12
D7S820: 8,10
THO1: 8,9
TPOX: 8,10
vWA: 16

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 0.1% Lactalbumin hydrolysate (LH) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service